Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyloxonium tetrafluoroborate, 95%

Catalog No. AC436761000
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 368-39-8 | C6H15BF4O | 189.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkyl transfer reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-39-8, 60-29-7
Molecular Formula C6H15BF4O
Molecular Weight (g/mol) 189.99
MDL Number MFCD00044423
InChI Key IYDQMLLDOVRSJJ-UHFFFAOYSA-N
Synonym triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium
PubChem CID 2723982
IUPAC Name triethyloxidanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CC[O+](CC)CC

Specifications

Melting Point 84.0°C to 89.0°C
Color White
Assay Percent Range 95%
Packaging Glass bottle
Quantity 100 g
Merck Index 15,5867
Solubility Information Solubility in water: decomposes
Stabilizer 3 to 5 % ether
Formula Weight 189.99
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Triethyloxonium tetrafluoroborate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Flammable solid.
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-705-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.