Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 8-Bromo-1-octene

Catalog No. 2523011G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 g
5 g

Catalog No. 25-230-11G

Supplier: Sigma Aldrich 2523011G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 8-bromo-1-octene has been used in preparation of polymerizable ligand, required for the synthesis of quantum dot-labelled polymer beads
  • Grignard reagent derived from 8-bromo-1-octene has been used in the synthesis of (2s,3s,5r)-5-[(1r)-1-hydroxy-9-decenyl]-2-pentyltetrahydro-3-furanol
  • It has been used as building block in natural product synthesis

Specifications

CAS 2695-48-9
Density 1.139 g/mL (at 25°C (literature))
Molecular Formula C8H15Br
Refractive Index n20/D 1.467 (literature)
Linear Formula Br(CH2)6 CH=CH2
Quantity 1 g
MDL Number MFCD00000275
Molecular Weight (g/mol) 191.11
Percent Purity 97%
Biological Activity Respiratory System
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.