Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Bromoadenine

Catalog No. p-7043111
Click to view available options
Quantity:
100 mg
500 mg
1 g

CAS: 6974-78-3 | C5H4BrN5 | 214.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6974-78-3
Molecular Formula C5H4BrN5
Molecular Weight (g/mol) 214.03
MDL Number MFCD00082518
InChI Key FVXHPCVBOXMRJP-UHFFFAOYSA-N
Synonym 8-bromoadenine, 8-bromo-9h-purin-6-amine, 6-amino-8-bromopurine, 8-br-adenine, 8-bromo-1h-adenine, 8-bromo-1h-purin-6-amine, 8-bromo-9h-purin-6-ylamine, 8-bromopurine-6-ylamine, chembl65747, 9h-purin-6-amine, 8-bromo
PubChem CID 81457
IUPAC Name 8-bromo-7H-purin-6-amine
SMILES NC1=C2NC(Br)=NC2=NC=N1

Specifications

Melting Point >250°C
Color Brown to White
Quantity 100 mg
Formula Weight 214.03
Physical Form Powder
Chemical Name or Material 8-Bromoadenine
EINECSNumber 230-225-1
RTECSNumber UO7410000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.