Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Bromooctanoic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
17696-11-6
C8H15BrO2
223.11
MFCD00004430
BKJFDZSBZWHRNH-UHFFFAOYSA-N
8-bromocaprylic acid,8-bromo octanoic acid,octanoic acid, 8-bromo,8-bromo-n-octanoic acid,omega-bromocaprylic acid,8-bromo-octanoic acid,octanoicacid, 8-bromo,acmc-1bo24,8-bromooctanoic acid,ksc180g8h
548275
8-bromooctanoic acid
C(CCCC(=O)O)CCCBr
17696-11-6
C8H15BrO2
223.11
MFCD00004430
BKJFDZSBZWHRNH-UHFFFAOYSA-N
8-bromocaprylic acid,8-bromo octanoic acid,octanoic acid, 8-bromo,8-bromo-n-octanoic acid,omega-bromocaprylic acid,8-bromo-octanoic acid,octanoicacid, 8-bromo,acmc-1bo24,8-bromooctanoic acid,ksc180g8h
548275
8-bromooctanoic acid
C(CCCC(=O)O)CCCBr
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point >110°C
Quantity 50g
Melting Point 36.0°C to 41.0°C
Boiling Point 147.0°C to 150.0°C (2.0mmHg)
Color White to Cream
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Beilstein 02, IV, 1000
Formula Weight 223.11
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material 8-Bromooctanoic acid, 97%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger