Learn More
8-Bromooctanoic acid, 95%
Description
8-Bromooctanoic acid is used in the synthesis of 8-(N-Methyl-4,4'-bipyridinyl)- octanoic acid. 8-Mercaptooctanoic acid was prepared from 8-bromooctanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
8-Bromooctanoic acid is used in the synthesis of 8-(N-Methyl-4,4′-bipyridinyl)- octanoic acid. 8-Mercaptooctanoic acid was prepared from 8-bromooctanoic acid.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, bases.
Specifications
Specifications
| Melting Point | 37°C to 41°C |
| Boiling Point | 147°C to 150°C (2 mmHg) |
| Flash Point | >110°C (230°F) |
| Linear Formula | BrCH2(CH2)6CO2H |
| Quantity | 10 g |
| UN Number | UN1759 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 223.11 |
| Percent Purity | 97% |
| Chemical Name or Material | 8-Bromooctanoic acid |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.