Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Bromoquinoline, 98%, Thermo Scientific Chemicals

Catalog No. p-7028573
Click to view available options
Quantity:
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16567-18-3
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD00191859
InChI Key PIWNKSHCLTZKSZ-UHFFFAOYSA-N
Synonym quinoline, 8-bromo, 8-bromo-quinoline, 8-bromoquinoline, pubchem5816, 8-bromquinoline, acmc-209dt3, ksc181q5t, buttpark 33\04-57
PubChem CID 140109
IUPAC Name 8-bromoquinoline
SMILES BrC1=C2N=CC=CC2=CC=C1

Specifications

Density 1.594
Boiling Point 302°C to 304°C
Flash Point >110°C (230°F)
Refractive Index 1.6725
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 208.06
Percent Purity 98%
Chemical Name or Material 8-Bromoquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.