Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Chloro-1-octanol, 98%

Catalog No. AC374900250
Click to view available options
Quantity:
25 mL

CAS: 23144-52-7 | C8H17ClO | 164.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23144-52-7
Molecular Formula C8H17ClO
Molecular Weight (g/mol) 164.67
InChI Key YDFAJMDFCCJZSI-UHFFFAOYSA-N
Synonym 8-chloro-1-octanol, 1-octanol, 8-chloro, 8-chlorooctanol, 8-chloro-octan-1-ol, 8-chloro-1-n-octanol, 8-chlor-1-octanol, 8-chlor-octan-1-ol, acmc-1cr4x
PubChem CID 90012
IUPAC Name 8-chlorooctan-1-ol
SMILES C(CCCCCl)CCCO

Specifications

Density 0.9760g/mL
Boiling Point 129.0°C to 130.0°C (15.0 mmHg)
Flash Point >113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4575 to 1.4595
Linear Formula Cl(CH2)8OH
Quantity 25 mL
Specific Gravity 0.976
Formula Weight 164.67
Percent Purity 98%
Chemical Name or Material 8-Chloro-1-octanol
Show More Show Less
EINECSNumber 245-451-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.