Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Chloroquinoline 98.0+%, TCI America™
SDP

Catalog No. C028425G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 611-33-6
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.60
MDL Number MFCD00047618
InChI Key RUSMDMDNFUYZTM-UHFFFAOYSA-N
Synonym quinoline, 8-chloro, ccris 3982, 8-choloroquinoline, 8-chloro-quinoline, pubchem10742, acmc-1aumj, ksc163s9l, 8-chloro-quinolin;quinoline, 8-chloro
PubChem CID 69139
ChEBI CHEBI:48985
IUPAC Name 8-chloroquinoline
SMILES ClC1=C2N=CC=CC2=CC=C1

Specifications

Color Yellow
Boiling Point 288°C
Quantity 25 g
Formula Weight 163.60
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 8-Chloroquinoline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.