Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Ethoxyquinoline-5-sulfonic acid monohydrate, 99%

Catalog No. AAA1862218
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1862218 50 g
AAA1862209 10 g
AAA1862230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1862218 Supplier Thermo Scientific Chemicals Supplier No. A1862218
Add to Cart

CAS: 15301-40-3 | C11H11NO4S | 253.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15301-40-3
Molecular Formula C11H11NO4S
Molecular Weight (g/mol) 253.27
MDL Number MFCD00065059
InChI Key YAMVZYRZAMBCED-UHFFFAOYSA-N
Synonym actinoquinol, 8-ethoxy-5-quinolinesulfonic acid, 5-quinolinesulfonic acid, 8-ethoxy, unii-470vqe23o3, 8-ethoxy-5-chinolinsulfonsaeure, actinochinolum, actinoquinolum, actinoquinol inn, actinoquinolum inn-latin, sodium tequinol
PubChem CID 23674
IUPAC Name 8-ethoxyquinoline-5-sulfonic acid
SMILES CCOC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O

Specifications

Melting Point ∼296°C (decomposition)
Quantity 50 g
Beilstein 225878
Formula Weight 271.29 (253.28 Anhydrous)
Percent Purity 99%
Chemical Name or Material 8-Ethoxyquinoline-5-sulfonic acid monohydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-334-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.