Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Hydroxy-7-propylquinoline 98.0+%, TCI America™
SDP

Catalog No. P05311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P05311G 1 g
P05315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P05311G Supplier TCI America Supplier No. P05311G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58327-60-9
Molecular Formula C12H13NO
Molecular Weight (g/mol) 187.242
MDL Number MFCD00059755
InChI Key OKPSVYHYNXVYED-UHFFFAOYSA-N
Synonym 7-Propyloxine, 7-Propyl-8-quinolinol
PubChem CID 11604513
IUPAC Name 7-propylquinolin-8-ol
SMILES CCCC1=C(C2=C(C=CC=N2)C=C1)O

Specifications

Melting Point 0°C
Boiling Point 149°C
Quantity 1 g
Formula Weight 187.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 8-Hydroxy-7-propylquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.