Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
8-Hydroxyquinoline (Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Poly Bottle
Specifications
Specifications
| Alpha Vector | Hydroxyquinoline |
| Melting Point | 72°C |
| Color | White Beige Yellow |
| Boiling Point | 267°C |
| Ignition Residue | 0.05% max. |
| Packaging | Poly Bottle |
| Linear Formula | HOC6H3N:CHCH:CH |
| Quantity | 500 g |
| Solubility Information | 0.05% max. Insoluble in Alcohol |
| Vapor Pressure | 4.7hPa at 100°C |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Harmful by inhalation, in contact with skin and if swallowed. Possible risks of irreversible effects. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting. NFPA Health:2 Flammability:1 Instability:1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction