Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 8-Hydroxyquinoline copper(II) salt

Catalog No. 5507955G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
55-079-55G 5 g
1 options

Catalog No. 55-079-55G

Supplier: Sigma Aldrich 5507955G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

8-Hydroxyquinoline copper(II) salt

Specifications

CAS 13014-03-4
Melting Point 240°C
Molecular Formula C18H12CuN2O2
Linear Formula Cu(C9H6NO)2
Quantity 5 g
MDL Number MFCD00067392
Molecular Weight (g/mol) 351.85
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.