Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Hydroxyquinoline sulfate monohydrate 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic Bottle
207386-91-2
0·5 H2O
203.2
MFCD00149610
BNCXJZDIJIVJJO-UHFFFAOYSA-N
8-quinolinol hemisulfate,8-quinolinol hemisulfate hemihydrate,8-hydroxyquinoline hemisulfate salt hemihydrate,quinolin-8-ol; sulfuric acid; hydrate,quinolin-8-ol sulfate hydrate 2:1:1,8-hydroxyquinoline sulfate,8-quinolinol sulfate 2:1 monohydrate,8-quinolinol, sulfate, hydrate 2:1:1,8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard
207386-91-2
0·5 H2O
203.2
MFCD00149610
BNCXJZDIJIVJJO-UHFFFAOYSA-N
8-quinolinol hemisulfate,8-quinolinol hemisulfate hemihydrate,8-hydroxyquinoline hemisulfate salt hemihydrate,quinolin-8-ol; sulfuric acid; hydrate,quinolin-8-ol sulfate hydrate 2:1:1,8-hydroxyquinoline sulfate,8-quinolinol sulfate 2:1 monohydrate,8-quinolinol, sulfate, hydrate 2:1:1,8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard
Packaging Glass bottle
Melting Point 182.0°C
Infrared Spectrum Authentic
Color Yellow
Water 4.5% max.
Quantity 100g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Merck Index 14, 4843
Solubility Information Solubility in water: soluble in water. Other solubilities: slightly soluble in alcohol, insoluble in ether
Formula Weight 203.2
Physical Form Crystalline Powder or Lumps
Percent Purity 98%
Chemical Name or Material 8-Hydroxyquinoline sulfate monohydrate, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning