Learn More
Description
- Made of glazed porcelain
- Designed with one long, flat end and knobbed handle for secure grip
Specifications
Specifications
| Quantity | 1 Spatula |
| Product Type | Spatula |
| Length (English) | 8 in. |
| Material | Porcelain |
| Length (Metric) | 20 cm |
| Autoclavable | Autoclavable |
| Shape | Flat End |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

