Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Methoxypsoralen, MilliporeSigma™ Supelco™

Catalog No. p-7187765 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 g
5 g
Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 298-81-7
Physical Form Crystals
Linear Formula C12H8O4
MDL Number MFCD00005009
Melting Point 148°C to 150°C
Synonym 8-MOP; 9-Methoxyfuro[3,2-g][1 ]benzopyran-7-one; Ammoidin; Methoxsalen; Xanthotoxin
Solubility Soluble in acetic acid
Type Analytical Standard
Quantity 1 g
Formula Weight 216.19
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.