Learn More
8-Nitroquinoline, 98%
Description
8-Nitroquinoline is used in the preparation of bisubstituted phenoxy-6-methoxy-8-aminoquinoline. It is used to prepare furazano [3,4-h] quinoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
8-Nitroquinoline is used in the preparation of bisubstituted phenoxy-6-methoxy-8-aminoquinoline. It is used to prepare furazano [3,4-h] quinoline.
Solubility
Soluble in ethanol, ethyl ether and benzene, slightly soluble in cold water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications
Specifications
| Melting Point | 86°C to 90°C |
| Quantity | 5 g |
| Beilstein | 135178 |
| Solubility Information | Soluble in ethanol,ethyl ether and benzene,slightly soluble in cold water. |
| Formula Weight | 174.16 |
| Percent Purity | 98% |
| Chemical Name or Material | 8-Nitroquinoline |
Safety and Handling
| Hazard Category | H315-H319-H335-H341-H351 |
| Hazard Statement | GHS H Statement H341-H351-H315-H319-H335 Suspected of causing genetic defects. Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c |
| EINECSNumber | 210-135-9 |
| RTECSNumber | VC1925000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.