Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-oxo-dG Antibody, R&D Systems™
GSA_VAGREENER_CHOICESDP

Catalog No. 4354MC050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4354MC050 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4354MC050 Supplier R&D Systems Supplier No. 4354MC050

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 38 publications

8-oxo-dG Monoclonal specifically detects 8-oxo-dG in Drosophila∣Human∣Mouse∣Multi-species∣Rat∣Transgenic Mouse samples. It is validated for Bioassay∣ChIP Assay∣Co-immunoprecipitation (Co-IP)∣Comet Assay∣ELISA∣Flow Cytometry∣Immunocytochemistry∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Western Blot.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Antigen 8-oxo-dG
Applications Bioassay, ChIP Assay, Co-immunoprecipitation (Co-IP), Comet Assay, ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 15A3
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:250 dilution, Immunocytochemistry 1:250 dilution
Formulation This antibody is provided as purified immunoglobulin from mouse ascites at 0.5 mg/ml in 1X PBS containing 0.1% sodium azide, 50% glycerol. with 0.1% Sodium Azide
Host Species Mouse
Immunogen 8-oxo-dG-conjugated-KLH
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This mouse monoclonal antibody specifically binds to 8-hydroxy-2'- deoxyguanosine within DNA in H 2 O 2 -treated cells.
Target Species Drosophila, Human, Mouse, Multi-species, Rat, Transgenic Mouse
Content And Storage Store the unopened product at -20 to -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.