Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Amplitube™ Strips of 8 Caps

Catalog No. 23045240 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Natural
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-240 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-045-240 Supplier Simport™ Scientific Supplier No. T3211N
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amplitube™ Strips of 8 Caps to be used with T320-1 tube strip series. They ensure a perfect closure and a positive seal during thermal cycling and prevent evaporation.

  • Made of polypropylene
  • RNase, DNase, Pyrogen and DNA-free
  • Autoclavable
  • Nonsterile

Specifications

Certifications/Compliance RNase, DNase, Pyrogen and DNA-free
Pre-sterilized No
Product Type Domed Strip Cap
Autoclavable Autoclavable
Material Polypropylene
Color Natural
Sterility Non-sterile
For Use With (Equipment) T320-1 Series Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.