Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

80 Octane Blend, PRF, B&J Brand, Meets requirements of ASTM D2699 and D2700, Honeywell Burdick & Jackson

Catalog No. p-7112074 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
19 L
204 L
Packaging:
Steel Can
Steel Drum

Specifications

Chemical Name or Material 80 Octane Blend, PRF
Name Note B&J Brand, Meets requirements of ASTM D2699 and D2700
CAS 540-84-1, 142-82-5
Boiling Point 100°C
CAS Max % 80.0000%, 20.0000%
UN Number UN1993
Density 0.6859 g/cm3
Grade Primary Reference Fuel
Packaging Steel Can
Quantity 19 L
Flash Point −12.2°C
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.