Learn More
Moldex™ 8000 Series Half-Mask Respirators Cartridges
SDP
Catalog No. p-57182 Shop All Moldex Metric Inc. Products
Click to view available options
Cartridge Type:
Organic Vapor Cartridge
Organic Vapor, Acid Gas Cartridge
Particulate
For Use With (Application):
For protection against asbestos, lead, inorganic arsenic, cadmium, for use in environments where oil based particulates are present
For protection against organic vapors
For protection against organic vapors, chlorine, hydrogen chloride, sulfur dioxide, chlorine dioxide, hydrogen sulfide and hydrogen fluoride
This item is not returnable. View return policy
This item is not returnable. View return policy
For use with 8000 series half mask respirators
  • Use with 8910 N95, 8970 R95 or 8940 P100 filters against particulates
  • The P100 cartridge can be used for both oil and non-oil based particulates
  • 100% PVC-Free™
Specifications
Cartridge Type Organic Vapor Cartridge
Certifications/Compliance NIOSH
Product Type Organic Vapor Cartridge
For Use With (Application) For protection against organic vapors
For Use With (Equipment) Series 8000 half mask, n95, r95, p100 filter
Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.