Learn More
Moldex™ 8000 Series Paint Spray/Pesticide Pre-Assembled Respirators
SDP
Pre-assembled half-mask respirators equipped for oil-free paint spray and pesticide work environments. Kit includes 8000 Series facepiece, 8100 Organic Vapor Cartridges, 8910 N95 Pre-Filters and 8020 Pre-Filter Retainers.
Specifications
Certifications (NIOSH) NIOSH 42 CFR Part 84, TC-23C-1223
Color Gray
Filter Type N95
Material Thermoplastic Elastomer
Product Type Pre-Assembled Half-Mask Respirator
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-003-343A
SDP
View Documents
Moldex™
8111N
Small N/A N/A N/A
19-003-343B
SDP
View Documents
Moldex™
8112N
Medium N/A N/A N/A
Description
Description
  • 8000 Series half-mask pre-assembled with OV cartridges, N95 pre-filters and pre-filter retainers
  • For oil-free paint sprays and pesticides
  • Soft, durable facepiece and lightweight design with extra-wide sealing
  • Exhalation valve positioned for better communication
  • Cartridges have a snap-in mount for easy replacement and no misthreading
  • Living hinge allows head harness to be folded comfortably around neck when not in use
  • Low profile, compact design fits well under welding helmets and with safety glasses
  • Maintenance made simple with very few replacement parts
Specifications
Specifications
NIOSH 42 CFR Part 84, TC-23C-1223
N95
Pre-Assembled Half-Mask Respirator
8000 Facepiece, 8100 Organic Vapor Cartridges, 8910 N95 Pre-Filters and 8020 Pre-Filter Retainers
Gray
Thermoplastic Elastomer
Paint spray and pesticide applications which are oil-free
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.