Learn More
Thermo Scientific™ 81-Well Cryogenic Storage Racks with Lid
GREENER_CHOICE
Catalog No. p-4520568
Click to view available options
Color:
Fluorescent Green
Fluorescent Orange
Fluorescent Pink
Natural
Hold eight-one 1.5 or 2.0mL cryogenic storage vials with this stackable cryogenic storage rack with lid.
Hold eighty-one 1.5 or 2.0mL cryogenic storage vials with the Thermo Scientific™ 81-Well Cryogenic Storage Rack with Lid. Available in several colors with transparent tinted lid that protects samples from dust and debris and permits stacking.
  • Molded with alphanumeric grid for easy tube identification
  • Lid protects samples and allows racks to be stacked
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Holds 81 (1.5 or 2.0 mL Tubes)
Material Polypropylene
Type 81-Well Cryogenic Storage Racks with Lid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.