Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ mutaFISH™ DNA Accessory Kit 1 for Cells

Catalog No. 89333042 Shop All Abnova Corporation Products
Click to view available options
Quantity:
1 Kit

mutaFISH™ DNA Accessory Kit 1 for Cells provides reagents and enzymes for in situ restriction digestion, exonucleolysis, mutaFISH™ hybridization, ligation and amplification in cell prior to mutaFISH™.

Includes:

Box 1

  • H20
  • PBS
  • Wash Buffer 1
  • Digestion Enzyme Buffer
  • Wash Buffer 2
  • Formamide
  • Detection Buffer

Box 2

  • Digestion Enzyme
  • BSA
  • Restriction Enzyme 1
  • 10X Restriction Enzyme Buffer
  • Hybrid Enzyme
  • 10X Hybrid Enzyme Buffer
  • 1M KCL
  • DNA Polymerase
  • 10X DNA Pol Buffer
  • dNTP Mix
  • 50% Glycerol
  • DAPI (100μM)

Specifications

Content And Storage Store Box 1 at 4°C, store Box 2 at -20°C.
Quantity 1 Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.