Learn More
MCR Safety NitriShield™ Nitrile Disposable Gloves
Catalog No. 19812758 Shop All MCR Safety Products
Click to view available options
Size:
2X-Large
Large
Medium
Small
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
8 mil Nitrile industrial and food service grade gloves
  • Nitrile gloves contain no latex proteins and offer excellent resistance to punctures and tears
  • Nitrile gloves are three times more puncture resistant than rubber and can be used to offer superior resistance to many types of chemicals
  • Textured grip
  • This glove is an excellent alternative for individuals who experience allergic reactions to latex
  • The 6030 is an industrial / food service grade Powder free nitrile glove that measures 8 mil in thickness and is 9.5 inches in length
  • Component materials comply with all federal regulations for food contact
  • Specifications
    Certifications/Compliance FDA Compliant
    Color Blue
    Disposable Single-use
    Exterior Finish Partially Textured
    Hand Ambidextrous
    Latex Free Yes
    Material Nitrile
    Size 2X-Large
    Thickness (English) 8 mil
    Thickness (Metric) 0.20 mm
    Quantity 50/Cs.
    Length (English) 9.5 in.
    Show More Show Less
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.