Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Bis(6-methoxy-2-naphthyl)anthracene 98.0+%, TCI America™
SDP

Catalog No. B50461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50461G 1 g
B5046200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50461G Supplier TCI America Supplier No. B50461G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 235099-48-6
Molecular Formula C36H26O2
Molecular Weight (g/mol) 490.602
MDL Number MFCD12022382
InChI Key VEVBQRGODPIBNJ-UHFFFAOYSA-N
PubChem CID 18710993
IUPAC Name 9,10-bis(6-methoxynaphthalen-2-yl)anthracene
SMILES COC1=CC2=C(C=C1)C=C(C=C2)C3=C4C=CC=CC4=C(C5=CC=CC=C53)C6=CC7=C(C=C6)C=C(C=C7)OC

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 490.60
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Bis(6-methoxy-2-naphthyl)anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.