Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Bis(chloromethyl)anthracene 97.0+%, TCI America™
SDP

Catalog No. B132725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B132725G 25 g
B13275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B132725G Supplier TCI America Supplier No. B132725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10387-13-0
Molecular Formula C16H12Cl2
Molecular Weight (g/mol) 275.172
MDL Number MFCD00045388
InChI Key UOSROERWQJTVNU-UHFFFAOYSA-N
PubChem CID 25219
IUPAC Name 9,10-bis(chloromethyl)anthracene
SMILES C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2CCl)CCl

Specifications

Quantity 25 g
UN Number 3261
Formula Weight 275.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Bis(chloromethyl)anthracene
RTECSNumber CA9365000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.