Learn More
9,10-Bis(phenylethynyl)anthracene, Thermo Scientific Chemicals
Description
9,10-Bis(phenylethynyl)anthracene is used as a chemiluminescent fluorophore with high quantum efficiency. It is used in lightsticks as a fluorophor producing ghostly green light. It is also used as a dopant for organic semiconductors in OLEDs. It is used as a reagent for chemiluminescence research. 9,10-Bis(phenylethynyl)anthracene is known to exert fluorescence properties due to its polycyclic aromatic hydrocarbon system and it is commonly used as an scintillator additive. 9,10-Bis(phenylethynyl)anthracene was also studied extensively as a fluorescence emitters for peroxyoxalate chemiluminescence.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 254°C to 255°C |
Quantity | 5 g |
Beilstein | 1891432 |
Sensitivity | Light sensitive |
Solubility Information | It is insoluble in water but is soluble in most organic solvents such as carbon disulfide,alcohols,benzene,chloroform,and hydronaphthalenes. |
Formula Weight | 378.47 |
Physical Form | Crystalline |
Chemical Name or Material | 9,10-Bis (phenylethynyl)anthracene |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 233-210-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.