Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Dibromo-2-methylanthracene 96.0+%, TCI America™
SDP

Catalog No. D49771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49771G 1 g
D4977200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49771G Supplier TCI America Supplier No. D49771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 177839-45-1
Molecular Formula C15H10Br2
Molecular Weight (g/mol) 350.05
MDL Number MFCD00266492
InChI Key PNDNWMMKRSTYTC-UHFFFAOYSA-N
PubChem CID 1713096
IUPAC Name 9,10-dibromo-2-methylanthracene
SMILES CC1=CC=C2C(Br)=C3C=CC=CC3=C(Br)C2=C1

Specifications

Melting Point 143°C
Color Yellow
Quantity 1 g
Formula Weight 350.05
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Dibromo-2-methylanthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.