Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Dibromoanthracene, 98%

Catalog No. p-7045983
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 523-27-3 | C14H8Br2 | 336.026 g/mol

9,10-Dibromoanthracene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Dibromoanthracene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 523-27-3
Molecular Formula C14H8Br2
Molecular Weight (g/mol) 336.026
MDL Number MFCD00001244
InChI Key BRUOAURMAFDGLP-UHFFFAOYSA-N
Synonym anthracene, 9,10-dibromo, 9,10-dibromo-anthracene, unii-61cp7c5y82, 9,10-dibromanthracen, 9,10-dibromo anthracenen, pubchem15115, 9,10 dibromoanthracene, anthracene,10-dibromo, 9,10-dibromoanthracene, acmc-1az4y
PubChem CID 68226
IUPAC Name 9,10-dibromoanthracene
SMILES C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2Br)Br

Specifications

Melting Point 222°C to 223°C (sublimation)
Quantity 5 g
Beilstein 1912109
Merck Index 14,3018
Solubility Information Soluble in water.
Formula Weight 336.04
Percent Purity 98%
Chemical Name or Material 9,10-Dibromoanthracene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-342-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.