Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Dichloroanthracene 96.0+%, TCI America™
SDP

Catalog No. D032710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D032710G 10 g
D03271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D032710G Supplier TCI America Supplier No. D032710G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 605-48-1
Molecular Formula C14H8Cl2
Molecular Weight (g/mol) 247.118
MDL Number MFCD00001246
InChI Key FKDIWXZNKAZCBY-UHFFFAOYSA-N
Synonym anthracene, 9,10-dichloro, acmc-209mkk, anthracene,10-dichloro, 9,10-dichloro-anthracene, anthracene,9,10-dichloro, 9.10-dichloroanthracene
PubChem CID 11800
IUPAC Name 9,10-dichloroanthracene
SMILES C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2Cl)Cl

Specifications

Melting Point 210°C
Quantity 10 g
Formula Weight 247.12
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Dichloroanthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.