Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Dimethylanthracene, 97%

Catalog No. p-7026456
Click to view available options
Quantity:
1 g
5 g

CAS: 781-43-1 | C16H14 | 206.29 g/mol

9,10-Dimethylanthracene is used as an antioxidant-photosensitizer dual-loaded polymeric micelles with controllable production of reactive oxygen species. It is also used as chemical rate constant actinometer in singlet molecular oxygen reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Dimethylanthracene is used as an antioxidant-photosensitizer dual-loaded polymeric micelles with controllable production of reactive oxygen species. It is also used as chemical rate constant actinometer in singlet molecular oxygen reactions.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 781-43-1
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00001262
InChI Key JTGMTYWYUZDRBK-UHFFFAOYSA-N
Synonym anthracene, 9,10-dimethyl, unii-di986077vr, ccris 2742, 9,10-dimethyl-anthracene, gnf-pf-2329, acmc-209pdj, 9,10-dimethylanthracen, 9:10-dimethylanthracene, anthracene,10-dimethyl, 9,10-dimethylanthracene
PubChem CID 13076
IUPAC Name 9,10-dimethylanthracene
SMILES CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12

Specifications

Melting Point 182°C to 184°C
Quantity 1 g
Beilstein 1909028
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 206.29
Percent Purity 99%
Chemical Name or Material 9,10-Dimethylanthracene
Hazard Category Warning
Hazard Statement GHS H Statement
H341-H351
Suspected of causing genetic defects.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 212-308-4
RTECSNumber CA9685000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.