Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Dimethylanthracene 98.0+%, TCI America™
SDP

Catalog No. D025210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D025210G 10 g
D02521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D025210G Supplier TCI America Supplier No. D025210G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 781-43-1
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00001262
InChI Key JTGMTYWYUZDRBK-UHFFFAOYSA-N
Synonym anthracene, 9,10-dimethyl, unii-di986077vr, ccris 2742, 9,10-dimethyl-anthracene, gnf-pf-2329, acmc-209pdj, 9,10-dimethylanthracen, 9:10-dimethylanthracene, anthracene,10-dimethyl, 9,10-dimethylanthracene
PubChem CID 13076
IUPAC Name 9,10-dimethylanthracene
SMILES CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12

Specifications

Melting Point 182°C
Color Yellow
Quantity 10 g
Formula Weight 206.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Dimethylanthracene
EINECSNumber (4)-0640
RTECSNumber CA9685000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.