Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Diphenylanthracene, 99%

Catalog No. p-7022799
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 1499-10-1 | C26H18 | 330.43 g/mol

9,10-Diphenylanthracene acts as a sensitizer in chemiluminescence. It is used in light sticks in order to produce blue light. It is also useful in blue organic light-emitting diodes (OLEDs) and OLED-based displays. Further, it plays an important role in the preparation of 9,10-difluoro-9,10-diphenyl-9,10-dihydro-anthracene. In addition to this, it is used as an organic semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Diphenylanthracene acts as a sensitizer in chemiluminescence. It is used in light sticks in order to produce blue light. It is also useful in blue organic light-emitting diodes (OLEDs) and OLED-based displays. Further, it plays an important role in the preparation of 9,10-difluoro-9,10-diphenyl-9,10-dihydro-anthracene. In addition to this, it is used as an organic semiconductor.

Solubility
Soluble in ethanol, chloroform, benzene and carbon disulfide. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1499-10-1
Molecular Formula C26H18
Molecular Weight (g/mol) 330.43
MDL Number MFCD00001253
InChI Key FCNCGHJSNVOIKE-UHFFFAOYSA-N
Synonym anthracene, 9,10-diphenyl, 9,10-diphenyl anthracene, unii-51bq8iyq9u, 51bq8iyq9u, acmc-1brsc, 9, 9-diphenylanthracene, anthracene,10-diphenyl, 9,10-diphenylanthracene, 9,10-diphenylanthracene 1g, 9,10-diphenylanthracene, analytical standard
PubChem CID 15159
ChEBI CHEBI:51676
IUPAC Name 9,10-diphenylanthracene
SMILES C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12

Specifications

Melting Point 248°C to 254°C
Odor Odorless
Quantity 1 g
Beilstein 1914010
Solubility Information Soluble in ethanol,chloroform,benzene and carbon disulfide. Insoluble in water.
Formula Weight 330.43
Percent Purity 99%
Chemical Name or Material 9,10-Diphenylanthracene
EINECSNumber 216-105-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.