Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Phenanthrenequinone, 95%

Catalog No. aaa1176206
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 84-11-7 | C14H8O2 | 208.216 g/mol

9,10-Phenanthrenequinon may be used for high quality passivation on silicon (100) surfaces. Quinones may serve as substrates for a variety of flavoenzymes, useful fluorescent compound and quinone. Also used in dyes, as a hardener for dental restoration, for the determination of copper by spectrophotometric method, and for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Phenanthrenequinon may be used for high quality passivation on silicon (100) surfaces. Quinones may serve as substrates for a variety of flavoenzymes, useful fluorescent compound and quinone. Also used in dyes, as a hardener for dental restoration, for the determination of copper by spectrophotometric method, and for organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-11-7
Molecular Formula C14H8O2
Molecular Weight (g/mol) 208.216
MDL Number MFCD00001163
InChI Key YYVYAPXYZVYDHN-UHFFFAOYSA-N
Synonym 9,10-phenanthrenequinone, phenanthrenequinone, 9,10-phenanthrenedione, 9,10-phenanthraquinone, phenanthraquinone, 9,10-phenanthroquinone, 9-10 phenanthrene quinone, unii-42l7bz8h74, ccris 7615, phenanthrene, 9,10-dihydro-9,10-dioxo
PubChem CID 6763
ChEBI CHEBI:37454
IUPAC Name phenanthrene-9,10-dione
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C(=O)C2=O

Specifications

Melting Point 207°C to 212°C
Density 1.405
Boiling Point >360°C
Flash Point 245°C (473°F)
Quantity 5 g
UN Number UN3077
Beilstein 608838
Merck Index 14,7213
Solubility Information Insoluble in water.
Formula Weight 208.22
Percent Purity 95%
Chemical Name or Material 9,10-Phenanthrenequinone
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 201-515-5
RTECSNumber SF7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.