Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9(10H)-Acridone 98.0+%, TCI America™
SDP

Catalog No. A013325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A013325G 25 g
A01335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A013325G Supplier TCI America Supplier No. A013325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 578-95-0
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00005019
InChI Key FZEYVTFCMJSGMP-UHFFFAOYSA-N
Synonym acridone, 9 10h-acridone, acridin-9 10h-one, 9-acridone, 9 10h-acridinone, acridanone, 9-acridanone, acridin-9-one, 9-acridinol, 9,10-dihydro-9-oxoacridine
PubChem CID 2015
ChEBI CHEBI:50756
IUPAC Name 10H-acridin-9-one
SMILES C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3N2

Specifications

Melting Point 354°C
Color Yellow
Quantity 25 g
Formula Weight 195.22
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9(10H)-Acridone
RTECSNumber AR6976000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.