Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-(3,5-Dibromophenyl)carbazole 98.0+%, TCI America™
SDP

Catalog No. D49781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49781G 1 g
D4978200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49781G Supplier TCI America Supplier No. D49781G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 750573-26-3
Molecular Formula C18H11Br2N
Molecular Weight (g/mol) 401.101
MDL Number MFCD27939624
InChI Key YXHXRBIAIVNCPG-UHFFFAOYSA-N
PubChem CID 22667344
IUPAC Name 9-(3,5-dibromophenyl)carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC(=CC(=C4)Br)Br

Specifications

Melting Point 176°C
Color White
Quantity 1 g
Formula Weight 401.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(3,5-Dibromophenyl)carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.