Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9-(4-Bromophenyl)carbazole, 98%

Click to view available options
Quantity:
1g
5g
25g
57102-42-8
C18H12BrN
322.205
MFCD11617969
XSDKKRKTDZMKCH-UHFFFAOYSA-N
9-4-bromophenyl-9h-carbazole,9-4-bromophenyl carbazole,9-4-bromophenyl-9h-carbazole,9-4-bromo-phenyl-9h-carbazole,9h-carbazole, 9-4-bromophenyl,acmc-209lw0,n-4-bromophenyl carbazole,amth018,9-4-bromophenyl carbazole,9-4-bromophenyl carbazole
22361390
9-(4-bromophenyl)carbazole
C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)Br
57102-42-8
C18H12BrN
322.205
MFCD11617969
XSDKKRKTDZMKCH-UHFFFAOYSA-N
9-4-bromophenyl-9h-carbazole,9-4-bromophenyl carbazole,9-4-bromophenyl-9h-carbazole,9-4-bromo-phenyl-9h-carbazole,9h-carbazole, 9-4-bromophenyl,acmc-209lw0,n-4-bromophenyl carbazole,amth018,9-4-bromophenyl carbazole,9-4-bromophenyl carbazole
22361390
9-(4-bromophenyl)carbazole
C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)Br
Applications
It plays an important role as intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 5g
Melting Point 147°C to 151°C
Beilstein 199590
Formula Weight 322.2
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 9-(4-Bromophenyl)carbazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning