Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Bis[4-(1-pyrenyl)phenyl]fluorene 96.0+%, TCI America™
SDP

Catalog No. B49801G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49801G 1 g
B4980200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49801G Supplier TCI America Supplier No. B49801G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1174006-47-3
Molecular Formula C57H34
Molecular Weight (g/mol) 718.899
MDL Number MFCD12022454
InChI Key WHLKCPQPGTUZBU-UHFFFAOYSA-N
PubChem CID 101502333
IUPAC Name 1-[4-[9-(4-pyren-1-ylphenyl)fluoren-9-yl]phenyl]pyrene
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC=C(C=C4)C5=C6C=CC7=CC=CC8=C7C6=C(C=C8)C=C5)C9=CC=C(C=C9)C1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1

Specifications

Quantity 1 g
Formula Weight 718.90
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis[4-(1-pyrenyl)phenyl]fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.