Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Bis[4-(2-hydroxyethoxy)phenyl]fluorene 98.0+%, TCI America™
SDP

Catalog No. F044725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F044725G 25 g
F04475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F044725G Supplier TCI America Supplier No. F044725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 117344-32-8
Molecular Formula C29H26O4
Molecular Weight (g/mol) 438.52
MDL Number MFCD00216628
InChI Key NQXNYVAALXGLQT-UHFFFAOYSA-N
PubChem CID 3764404
IUPAC Name 2-(4-{9-[4-(2-hydroxyethoxy)phenyl]-9H-fluoren-9-yl}phenoxy)ethan-1-ol
SMILES OCCOC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OCCO)C=C1

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 25 g
Synonym 4,4′-(9-Fluorenylidene)bis(2-phenoxyethanol)
Formula Weight 438.52
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis[4-(2-hydroxyethoxy)phenyl]fluorene
EINECSNumber (4)-1718
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.