Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Bis(4-aminophenyl)fluorene 98.0+%, TCI America™
SDP

Catalog No. B1549100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1549100G 100 g
B154925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1549100G Supplier TCI America Supplier No. B1549100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15499-84-0
Molecular Formula C25H20N2
Molecular Weight (g/mol) 348.449
MDL Number MFCD00039156
InChI Key KIFDSGGWDIVQGN-UHFFFAOYSA-N
PubChem CID 631552
IUPAC Name 4-[9-(4-aminophenyl)fluoren-9-yl]aniline
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC=C(C=C4)N)C5=CC=C(C=C5)N

Specifications

Melting Point 236°C
Color White-Yellow
Quantity 100 g
Synonym 4,4′-(9-Fluorenylidene)dianiline
Formula Weight 348.45
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-aminophenyl)fluorene
EINECSNumber (4)-1619
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.