Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Di-n-octylfluorene-2-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog No. p-7034283
Click to view available options
Quantity:
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302554-81-0
Molecular Formula C35H53BO2
Molecular Weight (g/mol) 516.617
MDL Number MFCD16294553
InChI Key VOASJDUTCQKQLE-UHFFFAOYSA-N
Synonym 2-9,9-dioctyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9,9-di-n-octylfluorene-2-boronic acid pinacol ester, 2-9,9-dioctyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 3-hydroxy-2,3-dimethylbutan-2-yl hydrogen 9,9-dioctyl-9h-fluoren-2-ylboronate, amtb255, 2-9,9-dioctylfluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9,9-dioctylfluorene-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolyl-9,9-dioctylfluorene, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-dioctylfluorene, 2-9,9-di-n-octyl-2-fluorenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 11477856
IUPAC Name 2-(9,9-dioctylfluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C3(CCCCCCCC)CCCCCCCC

Specifications

Quantity 5 g
Beilstein 8661191
Formula Weight 516.61
Percent Purity 95%
Chemical Name or Material 9,9-Di-n-octylfluorene-2-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.