Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 98%

Catalog No. AC378060250
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 161265-03-8 | C39H32OP2 | 578.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161265-03-8
Molecular Formula C39H32OP2
Molecular Weight (g/mol) 578.63
MDL Number MFCD00233866
InChI Key CXNIUSPIQKWYAI-UHFFFAOYSA-N
Synonym xantphos, 4,5-bis diphenylphosphino-9,9-dimethylxanthene, 9,9-dimethyl-4,5-bis diphenylphosphino xanthene, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphine, dimethylbisdiphenylphosphinoxanthene, unii-nmu72mog9b, nmu72mog9b, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphane, 5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl-diphenylphosphane, 4,5-bis-diphenylphosphanyl-9,9-dimethyl-9h-xanthene
PubChem CID 636044
SMILES CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 226.0°C to 230.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 10268
Solubility Information Solubility in water: insoluble
IUPAC Name (5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl)-diphenylphosphane
Formula Weight 578.63
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material 9, 9-Dimethyl-4, 5-bis(diphenylphosphino)xanthene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.