Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Dimethylxanthene, 98+%, Thermo Scientific Chemicals

Catalog No. AAH2734003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2734003 1 g
AAH2734006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2734003 Supplier Thermo Scientific Chemicals Supplier No. H2734003
Add to Cart

9,9-Dimethylxanthene is suitable for use in the preparation of 9,9-dimethylxanthene-4,5-dicarboxylic acid. It may be used in the preparation of 9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene. It may be used in the preparation of 4,6-bis(dimesitylboryl)dibenzofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,9-Dimethylxanthene is suitable for use in the preparation of 9,9-dimethylxanthene-4,5-dicarboxylic acid. It may be used in the preparation of 9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene. It may be used in the preparation of 4,6-bis(dimesitylboryl)dibenzofuran.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19814-75-6
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.28
MDL Number MFCD00134434
InChI Key MTVNAPYHLASOSX-UHFFFAOYSA-N
Synonym 9,9-dimethyl-9h-xanthene, 9h-xanthene, 9,9-dimethyl, 9,9-dimethylxantene, 9,9-dimethyl xanthene, xanthene, 9,9-dimethyl, ksc182m0n, 9,9-dimethylxanthene, 9,9-dimethyl-9h-xanthene #
PubChem CID 606997
SMILES CC1(C)C2=CC=CC=C2OC2=CC=CC=C12

Specifications

Melting Point 40°C to 43°C
Boiling Point 114°C to 115°C (0.6 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Solubility Information Insoluble in water.
IUPAC Name 9,9-dimethylxanthene
Formula Weight 210.28
Percent Purity ≥98%
Chemical Name or Material 9,9-Dimethylxanthene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.