Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Dimethylxanthene 98.0+%, TCI America™
SDP

Catalog No. D40551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40551G 1 g
D40555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40551G Supplier TCI America Supplier No. D40551G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19814-75-6
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.28
MDL Number MFCD00134434
InChI Key MTVNAPYHLASOSX-UHFFFAOYSA-N
Synonym 9,9-dimethyl-9h-xanthene, 9h-xanthene, 9,9-dimethyl, 9,9-dimethylxantene, 9,9-dimethyl xanthene, xanthene, 9,9-dimethyl, ksc182m0n, 9,9-dimethylxanthene, 9,9-dimethyl-9h-xanthene #
PubChem CID 606997
IUPAC Name 9,9-dimethyl-9H-xanthene
SMILES CC1(C)C2=CC=CC=C2OC2=CC=CC=C12

Specifications

Melting Point 38°C
Color White-Yellow
Boiling Point 115°C
Quantity 1 g
Formula Weight 210.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 9,9-Dimethylxanthene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.