Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Anthracenemethanol, 98%

Catalog No. AC181870250
Click to view available options
Quantity:
5 g
25 g
250 g
Packaging:
Glass bottle

CAS: 1468-95-7 | C15H12O | 208.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1468-95-7
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00001264
InChI Key JCJNNHDZTLRSGN-UHFFFAOYSA-N
Synonym 9-anthracenemethanol, 9-hydroxymethylanthracene, 9-anthrylcarbinol, 9-methylolanthracene, 9-anthrylmethanol, 9-hydroxymethylantracene, 9-anthracene methanol, 9-hydroxymethyl anthracene, anthracene-9-carbinol, ccris 7297
PubChem CID 73848
IUPAC Name anthracen-9-ylmethanol
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 160.0°C to 164.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 208.26
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 9-Anthracenemethanol, 98%
Show More Show Less
EINECSNumber 215-998-5
RTECSNumber CB0577500
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.