Alfa Aesar™ 9-Anthraldehyde, 98%

Manufacturer: Alfa Aesar™ A1125814

Quantity 25g

 View more versions of this product

Catalog No. AAA1125814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 642-31-9
    Molecular Formula C15H10O
    Formula Weight 206.24
    MDL Number MFCD00001254
    Beilstein 639167
    Synonym 9-Anthracenecarboxaldehyde
    Chemical Name or Material 9-Anthraldehyde

    Specifications

    Formula Weight 206.24
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C15H10O
    Quantity 25g
    TSCA TSCA
    MDL Number MFCD00001254
    CAS 642-31-9
    Beilstein 639167
    Synonym 9-Anthracenecarboxaldehyde
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 102° to 106°C
    Chemical Name or Material 9-Anthraldehyde
    Assay 98%

    Description

    Applications
    9-Anthraldehyde is used to prepare asymmetrical tridentate Schiff base ligands, 2-(9-anthrylmethyl-ideneamino)-4-methyl-phenol and functionalized ligand like 2-(anthracen-9-ylidene)-4,5-bis(diphenylphosphino)-4-cyclopentene-1,3-dione through Knoevenangel condensation with the diphosphine ligand 4,5-bis(diphenylphosphino)-4-cyclopentene-1,3-dione. It is also used as an intermediate for dyes, pigment, active pharmaceutical ingredients and other organic compounds. Further, it is involved in the Diels Alder reaction with benzenediazonium-2-carboxylate.

    Solubility
    Soluble in toluene.

    Notes
    Air sensitive. Incompatible with strong bases and strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit