Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Benzylcarbazole 98.0+%, TCI America™
SDP

Catalog No. B355825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B355825G 25 g
B35585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B355825G Supplier TCI America Supplier No. B355825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19402-87-0
Molecular Formula C19H15N
Molecular Weight (g/mol) 257.34
MDL Number MFCD00218286
InChI Key HBAKJBGOHINNQM-UHFFFAOYSA-N
PubChem CID 146485
IUPAC Name 9-benzyl-9H-carbazole
SMILES C(N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=CC=C1

Specifications

Melting Point 119°C
Color White
Boiling Point 268°C
Quantity 25 g
Formula Weight 257.34
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-Benzylcarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.