Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Bromo-1-nonanol 95.0+%, TCI America™
SDP

Catalog No. B178525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B178525G 25 g
B17855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B178525G Supplier TCI America Supplier No. B178525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55362-80-6
Molecular Formula C9H19BrO
Molecular Weight (g/mol) 223.15
MDL Number MFCD00063348
InChI Key USJDOLXCPFASNV-UHFFFAOYSA-N
Synonym 9-bromo-1-nonanol, 1-nonanol, 9-bromo, 9-bromo-nonan-1-ol, nonamethylene bromohydrin, 9-bromnonan-1-ol, 9-bromononanol, 9-bromononyl alcohol, pubchem23429, 9-bromo nonanol, acmc-1axfq
PubChem CID 108700
IUPAC Name 9-bromononan-1-ol
SMILES OCCCCCCCCCBr

Specifications

Boiling Point 126°C
Quantity 25 g
Formula Weight 223.15
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 9-Bromo-1-nonanol
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.