Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9-Bromo-10-phenylanthracene, 98%

Click to view available options
Quantity:
250mg
1g
5g
23674-20-6
C20H13Br
333.228
MFCD00230983
WHGGVVHVBFMGSG-UHFFFAOYSA-N
10-bromo-9-phenylanthracene,anthracene, 9-bromo-10-phenyl,9-bromo-10-phenyl-anthracene,ksc201k3d,9-bromo-10-phenyl anthracene,9-bromo-10-phenyl anthracene,anthracene, 9-bromo-10-phenyl
4155836
9-bromo-10-phenylanthracene
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=CC=CC=C42)Br
23674-20-6
C20H13Br
333.228
MFCD00230983
WHGGVVHVBFMGSG-UHFFFAOYSA-N
10-bromo-9-phenylanthracene,anthracene, 9-bromo-10-phenyl,9-bromo-10-phenyl-anthracene,ksc201k3d,9-bromo-10-phenyl anthracene,9-bromo-10-phenyl anthracene,anthracene, 9-bromo-10-phenyl
4155836
9-bromo-10-phenylanthracene
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=CC=CC=C42)Br
Applications
It is used as an OLED material.

Solubility
Insoluble in water. Soluble in hot toluene, dimethylformamide.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Quantity 1g
Color Yellow
Melting Point 154°C to 158°C
Beilstein 2277064
Formula Weight 333.23
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 9-Bromo-10-phenylanthracene

GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning