Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
9-Bromoanthracene 95.0+%, TCI America™

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Melting Point | 101°C |
Quantity | 25 g |
Formula Weight | 257.13 |
Percent Purity | ≥95.0% (GC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 9-Bromoanthracene |
Safety and Handling
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction